Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AF251705 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_134158.1 | NP_598919.1 | 3 | 827 | ||
GenBank mRNA | AA175302.1 | - | 2 | 196 | ||
AB091767.1 | - | 3 | 558 | |||
AB091768.1 | - | 3 | 547 | |||
AB098476.1 | - | 3 | 494 | |||
AF251705.1 | - | 3 | 914 | |||
AK050008.1 | - | 3 | 555 | |||
AK079395.1 | - | 3 | 643 | |||
AK154808.1 | - | 3 | 547 | |||
AY048685.1 | - | 3 | 827 | |||
AY457050.1 | - | 3 | 494 | |||
BC006801.1 | - | 3 | 558 | 3152857 | ||
BC111820.1 | - | 3 | 493 |