Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ankrd6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001012450.1 | NP_001012453.1 | 3 | 566 | ||
NM_001012451.1 | NP_001012454.1 | 2 | 277 | |||
NM_080471.3 | NP_536719.2 | 2 | 406 | |||
GenBank mRNA | AK083265.1 | - | 1 | 1794 | ||
AK133551.1 | - | 1 | 293 | |||
AK135024.1 | - | 2 | 261 | |||
AK173070.1 | - | 2 | 317 | |||
AY026320.2 | - | 1 | 198 | |||
BC065177.1 | - | 3 | 516 | 6825961 | ||
CF531081.1 | - | 2 | 406 | |||
CF533090.1 | - | 2 | 292 |