Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Elmo2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_080287.2 | NP_525026.2 | 12 | 1045 | ||
NM_207706.1 | NP_997589.1 | 13 | 1081 | |||
GenBank mRNA | AB093301.1 | - | 3 | 1626 | ||
AF398884.1 | - | 11 | 944 | |||
AK032033.1 | - | 13 | 1054 | |||
AK033918.1 | - | 12 | 1239 | |||
AK038097.1 | - | 12 | 1034 | |||
AK038455.1 | - | 12 | 1151 | |||
AK045428.1 | - | 12 | 1046 | |||
AK047040.1 | - | 12 | 1007 | |||
AK050823.1 | - | 12 | 1008 | |||
AK053574.1 | - | 13 | 1045 | |||
AK129023.1 | - | 12 | 1045 | |||
BC023954.1 | - | 13 | 1066 | 5320429 | ||
BC033418.1 | - | 11 | 975 |