Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Elmo1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_080288.2 | NP_525027.1 | 18 | 1974 | ||
NM_198093.3 | NP_932761.1 | 3 | 497 | |||
GenBank mRNA | AF398883.1 | - | 17 | 1638 | ||
AK032085.1 | - | 3 | 407 | |||
AK053306.1 | - | 3 | 408 | |||
AK220253.1 | - | 18 | 1862 | |||
BC024727.1 | - | 3 | 394 | |||
BC031782.1 | - | 7 | 712 | 3990906 |