Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Caskin2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_080643.2 | NP_542374.2 | 11 | 1586 | ||
GenBank mRNA | AK129297.1 | - | 11 | 1624 | ||
BC053083.1 | - | 11 | 1586 | 5704642 |