Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sec63 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153055.3 | NP_694695.3 | 8 | 938 | ||
GenBank mRNA | AK036629.1 | - | 8 | 633 | ||
AK049737.1 | - | 6 | 3059 | |||
AK163813.1 | - | 1 | 824 | |||
AY024346.1 | - | 8 | 735 | |||
AY238936.1 | - | 8 | 708 | |||
BC019366.1 | - | 8 | 702 | 5066908 | ||
BC031846.1 | - | 8 | 702 | 5066822 | ||
BC059816.1 | - | 8 | 937 | |||
BC080810.1 | - | 7 | 2241 |