Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Il25 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_080729.3 | NP_542767.1 | 3 | 1016 | ||
GenBank mRNA | AF458060.1 | - | 2 | 940 |