Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Wdr5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_080848.2 | NP_543124.1 | 8 | 780 | ||
GenBank mRNA | AF416510.1 | - | 8 | 785 | ||
AK075937.1 | - | 8 | 723 | |||
BC008547.1 | - | 3 | 160 | |||
BC016103.1 | - | 8 | 715 | 4919781 | ||
BC025801.1 | - | 8 | 757 | 5132525 |