Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Asb13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178283.4 | NP_840068.1 | 5, 6 | 805 | ||
GenBank mRNA | AF403041.1 | - | 5, 6 | 805 | ||
AK078289.1 | - | 5, 6 | 761 | |||
AK133892.1 | - | 5, 6 | 729 | |||
BC018240.1 | - | 5, 6 | 759 | 4239996 |