Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Elovl6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_130450.2 | NP_569717.1 | 4, 5 | 516 | ||
GenBank mRNA | AB072039.1 | - | 4, 5 | 463 | ||
AF480860.1 | - | 4, 5 | 435 | |||
AK029029.1 | - | 4, 5 | 518 | |||
AK165847.1 | - | 3, 4 | 540 | |||
AK167373.1 | - | 4, 5 | 482 | |||
AY053453.1 | - | 4, 5 | 497 | |||
BC040097.1 | - | 4, 5 | 458 | |||
BC051041.1 | - | 4, 5 | 513 | 4012676 | ||
BC098492.1 | - | 3, 4 | 513 | 30091954 | ||
BC100576.1 | - | 4, 5 | 458 | 30097278 |