Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Stard4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133774.4 | NP_598535.1 | 5 | 539 | ||
GenBank mRNA | AF480297.1 | - | 5 | 394 | ||
AF480298.1 | - | 5 | 394 | |||
AK042347.1 | - | 5 | 543 | |||
AK042691.1 | - | 5 | 499 | |||
AK054501.1 | - | 5 | 540 | |||
AK078858.1 | - | 5 | 543 | |||
AK085331.1 | - | 5 | 543 | |||
AK165641.1 | - | 5 | 545 | |||
AK205539.1 | - | 2 | 177 | |||
BC005642.1 | - | 5 | 539 | 3709232 |