Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Stard6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001289648.1 | NP_001276577.1 | 9 | 896 | ||
NM_001289649.1 | NP_001276578.1 | 9 | 875 | |||
NM_001289650.1 | NP_001276579.1 | 9 | 910 | |||
NM_029019.4 | NP_083295.1 | 9 | 965 | |||
GenBank mRNA | AF480303.1 | - | 6 | 612 | ||
AK014759.1 | - | 9 | 876 | |||
AK016984.1 | - | 9 | 876 | |||
BC061022.1 | - | 7 | 648 | 6774873 | ||
BC087893.1 | - | 9 | 887 | 6774832 | ||
JF440953.1 | - | 7 | 684 | |||
JF440954.1 | - | 7 | 684 |