Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PAX4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006193.2 | NP_006184.2 | 1 | 315 | ||
XM_011516276.2 | XP_011514578.1 | 4 | 589 | |||
GenBank mRNA | AB008913.1 | - | 1 | 133 | ||
AF043978.1 | - | 1 | 315 | |||
BC074761.2 | - | 1 | 142 | 30915220 | ||
BC107149.2 | - | 1 | 259 | 40032097 | ||
BC107150.1 | - | 1 | 259 | 40032102 |