Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nphs2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_130456.3 | NP_569723.1 | 4 | 576 | ||
GenBank mRNA | AJ302048.1 | - | 4 | 523 | ||
AK052746.1 | - | 4 | 585 | |||
AY050309.1 | - | 4 | 599 | |||
BC067401.1 | - | 4 | 576 | 30470258 | ||
FJ715770.1 | - | 4 | 584 | |||
FJ715771.1 | - | 4 | 614 | |||
FJ715772.1 | - | 4 | 614 | |||
FJ715773.1 | - | 4 | 614 | |||
FJ715774.1 | - | 4 | 614 | |||
FJ715775.1 | - | 4 | 614 | |||
FJ715776.1 | - | 4 | 614 |