Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hpcal4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_174998.4 | NP_778163.1 | 4, 5 | 611 | ||
GenBank mRNA | AB079893.1 | - | 3, 4 | 403 | ||
AK032591.1 | - | 4, 5 | 611 | |||
BC138631.1 | - | 3, 4 | 446 | 8861651 | ||
BC138632.1 | - | 3, 4 | 446 | 8861652 |