Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Oxr1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001130166.1 | NP_001123638.1 | 12 | 2212 | ||
NM_130885.2 | NP_570955.1 | 10 | 1853 | |||
GenBank mRNA | AF324899.1 | - | 10 | 1852 | ||
AK158703.1 | - | 12 | 2194 |