Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Papln | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001205343.1 | NP_001192272.1 | 19 | 2299 | ||
NM_130887.3 | NP_570957.2 | 18 | 2233 | |||
GenBank mRNA | AF314171.1 | - | 18 | 2206 | ||
AK053198.1 | - | 19 | 2301 | |||
AK086767.1 | - | 18 | 2215 | |||
AK138980.1 | - | 17 | 3099 | |||
BC132475.1 | - | 18 | 2173 | 40130752 | ||
BC137854.1 | - | 18 | 2173 | 8860873 | ||
BC144975.1 | - | 19 | 2239 | 9053523 |