Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Capn8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_130890.2 | NP_570960.2 | 15, 16 | 1762 | ||
GenBank mRNA | AB050202.1 | - | 15, 16 | 1758 | ||
AB061518.1 | - | 15, 16 | 1708 | |||
BC125504.1 | - | 15, 16 | 1734 | 40130019 | ||
BC125508.1 | - | 15, 16 | 1734 | 40130023 | ||
BC145055.1 | - | 15, 16 | 1734 |