Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Znrf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001168621.1 | NP_001162092.1 | 3 | 1115 | ||
NM_001168622.1 | NP_001162093.1 | 2 | 1019 | |||
NM_001168623.1 | NP_001162094.1 | 3 | 314 | |||
NM_133206.3 | NP_573469.1 | 3 | 1115 | |||
GenBank mRNA | AF378525.1 | - | 3 | 820 | ||
AK030575.1 | - | 2 | 165 | |||
AK076756.1 | - | 3 | 262 | |||
AK078819.1 | - | 3 | 341 | |||
AK079338.1 | - | 3 | 1115 | |||
AK181114.1 | - | 1 | 73 | |||
AK189177.1 | - | 1 | 73 | |||
AK216619.1 | - | 3 | 241 | |||
BC006765.1 | - | 3 | 715 | 3710377 | ||
BC086770.1 | - | 3 | 913 | 5720382 | ||
BM950431.1 | - | 2 | 196 |