Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zfp287 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133208.2 | NP_573471.2 | 5 | 2576 | ||
GenBank mRNA | AF281141.1 | - | 5 | 2576 | ||
AK029572.1 | - | 5 | 2156 | |||
BC023790.1 | - | 3 | 1595 | |||
BC117876.1 | - | 5 | 2301 | 40095930 | ||
BC117877.1 | - | 5 | 2301 | 40095933 |