Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tlr7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290755.1 | NP_001277684.1 | 5 | 1772 | ||
NM_001290756.1 | NP_001277685.1 | 4 | 1691 | |||
NM_001290757.1 | NP_001277686.1 | 3 | 1659 | |||
NM_001290758.1 | NP_001277687.1 | 4 | 1704 | |||
NM_133211.4 | NP_573474.1 | 3 | 1623 | |||
GenBank mRNA | AF334942.1 | - | 3 | 1566 | ||
AF334943.1 | - | 3 | 1580 | |||
AK036549.1 | - | 3 | 1581 | |||
AK154906.1 | - | 5 | 1730 | |||
AK156908.1 | - | 3 | 1627 | |||
AY035889.1 | - | 2 | 1440 | |||
BC120596.1 | - | 2 | 1485 | 40129519 | ||
BC132385.1 | - | 3 | 1553 | 40130662 |