Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Xpnpep1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133216.3 | NP_573479.3 | 20 | 1913 | ||
GenBank mRNA | AK052094.1 | - | 12 | 1055 | ||
AK080423.1 | - | 9 | 661 | |||
AK146033.1 | - | 20 | 1893 | |||
AK149676.1 | - | 20 | 1852 | |||
AK158597.1 | - | 18 | 1870 | |||
AK166715.1 | - | 20 | 1813 | |||
AK169024.1 | - | 19 | 1739 | |||
AK177605.1 | - | 2 | 59 | |||
AK195402.1 | - | 2 | 73 | |||
AK197404.1 | - | 2 | 73 | |||
BC021534.1 | - | 11 | 848 | |||
BC065174.1 | - | 20 | 1801 | 6415314 |