Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Bco2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133217.3 | NP_573480.1 | 7 | 1019 | ||
GenBank mRNA | AJ290392.1 | - | 6 | 862 | ||
BC107006.2 | - | 6 | 862 | 40046913 | ||
BC107007.2 | - | 6 | 862 | 40046914 |