Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sgk3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001037759.1 | NP_001032848.1 | 17 | 1560 | ||
NM_133220.2 | NP_573483.1 | 18 | 1679 | |||
NM_177547.3 | NP_808215.2 | 17 | 1572 | |||
GenBank mRNA | AF312007.1 | - | 18 | 1621 | ||
AK028722.1 | - | 17 | 1915 | |||
AK030314.1 | - | 17 | 1561 | |||
AK031133.1 | - | 18 | 1660 | |||
AK031328.1 | - | 17 | 1501 | |||
AK080469.1 | - | 1 | 3308 | |||
AK143836.1 | - | 18 | 1726 | |||
AK146561.1 | - | 17 | 1515 | |||
AK151181.1 | - | 18 | 1624 | |||
AK151793.1 | - | 15 | 1222 | |||
AK151867.1 | - | 18 | 1656 | |||
AK151996.1 | - | 18 | 1823 | |||
AK166414.1 | - | 17 | 1503 | |||
AK166531.1 | - | 17 | 1521 | |||
AK171120.1 | - | 17 | 1548 | |||
AK171186.1 | - | 17 | 1548 | |||
AK171678.1 | - | 17 | 1532 | |||
AK171959.1 | - | 17 | 1609 | |||
AK172323.1 | - | 17 | 1522 | |||
AK186891.1 | - | 3 | 173 | |||
BC018363.1 | - | 15 | 1210 | 2631682 |