Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zfp87 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133228.3 | NP_573491.2 | 4 | 899 | ||
GenBank mRNA | AF242377.1 | - | 4 | 802 | ||
AK028465.1 | - | 1 | 455 | |||
AK030724.1 | - | 4 | 847 | |||
AK049711.1 | - | 4 | 819 | |||
BC031988.1 | - | 4 | 802 | 4017149 |