Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ripply3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133229.2 | NP_573492.2 | 4 | 413 | ||
GenBank mRNA | AB063284.1 | - | 4 | 415 | ||
AK041355.1 | - | 4 | 403 | |||
AK172430.1 | - | 4 | 406 | |||
BC060291.1 | - | 4 | 371 | 30135244 | ||
CF170896.1 | - | 4 | 413 |