Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rfxap | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133231.2 | NP_573494.1 | 2 | 634 | ||
GenBank mRNA | AF335512.1 | - | 2 | 614 | ||
AK077640.1 | - | 2 | 632 | |||
AK087489.1 | - | 2 | 635 | |||
AK144633.1 | - | 2 | 621 | |||
BC019935.1 | - | 2 | 597 | 5041039 | ||
BC047123.1 | - | 2 | 597 |