Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Glcci1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286728.1 | NP_001273657.1 | 5 | 513 | ||
NM_001286729.1 | NP_001273658.1 | 5 | 510 | |||
NM_133236.3 | NP_573499.2 | 5 | 1186 | |||
GenBank mRNA | AK029524.1 | - | 5 | 512 | ||
AY098636.1 | - | 5 | 1232 | |||
BC050802.1 | - | 5 | 551 | 6771009 | ||
DV652660.1 | - | 1 | 85 |