Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PBX1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001204961.1 | NP_001191890.1 | 6 | 1347 | ||
NM_001204963.1 | NP_001191892.1 | 6 | 1347 | |||
NM_002585.3 | NP_002576.1 | 6 | 1347 | |||
XM_005245228.3 | XP_005245285.1 | 7 | 1515 | |||
XM_005245229.3 | XP_005245286.1 | 7 | 1108 | |||
XM_011509590.1 | XP_011507892.1 | 7 | 1515 | |||
XM_011509591.2 | XP_011507893.1 | 7 | 1515 | |||
XM_011509592.1 | XP_011507894.1 | 7 | 1515 | |||
XM_017001395.1 | XP_016856884.1 | 6 | 1347 | |||
XM_017001396.1 | XP_016856885.1 | 6 | 1347 | |||
GenBank mRNA | AK291415.1 | - | 6 | 1029 | ||
AK299673.1 | - | 6 | 963 | |||
AK301540.1 | - | 5 | 692 | |||
BC101578.1 | - | 6 | 936 | 8069084 | ||
BC143435.1 | - | 5 | 862 | |||
BC143451.1 | - | 6 | 936 | |||
BT006705.1 | - | 6 | 884 | |||
M31170.1 | - | 4 | 620 | |||
M86546.1 | - | 6 | 1007 |