Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Crb1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133239.2 | NP_573502.2 | 7 | 2777 | ||
GenBank mRNA | AF406641.1 | - | 7 | 2742 | ||
AF492496.1 | - | 6 | 2618 | |||
BC053331.1 | - | 2 | 1020 |