Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PBX1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001204961.1 | NP_001191890.1 | 7, 8 | 1549 | ||
NM_002585.3 | NP_002576.1 | 8, 9 | 1662 | |||
XM_005245228.3 | XP_005245285.1 | 9, 10 | 1830 | |||
XM_005245229.3 | XP_005245286.1 | 9, 10 | 1423 | |||
XM_011509591.2 | XP_011507893.1 | 8, 9 | 1717 | |||
GenBank mRNA | AK291415.1 | - | 8, 9 | 1344 | ||
BC101578.1 | - | 8, 9 | 1251 | 8069084 | ||
BC143435.1 | - | 7, 8 | 1177 | |||
BC143451.1 | - | 7, 8 | 1138 | |||
BX647313.1 | - | 2, 3 | 2257 | |||
CR749446.1 | - | 2, 3 | 7547 | |||
M31170.1 | - | 6, 7 | 935 | |||
M86546.1 | - | 8, 9 | 1322 |