Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rtkn2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081346.1 | NP_001074815.1 | 12 | 1370 | ||
GenBank mRNA | AK030986.1 | - | 11 | 1251 | ||
AK045134.1 | - | 1268 | ||||
AK052662.1 | - | 11 | 1405 | |||
AK082859.1 | - | 4 | 408 | |||
AK087079.1 | - | 2 | 115 | |||
AK087239.1 | - | 3 | 274 | |||
BC116341.2 | - | 1369 | 40098130 | |||
BC116342.2 | - | 1369 | 40098131 | |||
BC158093.1 | - | 11 | 1372 | 9007380 |