Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ms4a3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133246.5 | NP_573509.1 | 4, 5 | 488 | ||
GenBank mRNA | AF280401.1 | - | 4, 5 | 427 | ||
AF321128.1 | - | 4, 5 | 427 | |||
AK036464.1 | - | 4, 5 | 490 | |||
AK158372.1 | - | 4, 5 | 489 | |||
AY184359.1 | - | 4, 5 | 442 | |||
BC117013.1 | - | 4, 5 | 404 | 40126332 | ||
BC117015.1 | - | 4, 5 | 404 | 40126334 | ||
BC144853.1 | - | 4, 5 | 404 | 9053401 |