Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Usp33 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001076676.2 | NP_001070144.1 | 13 | 1714 | ||
NM_001252486.1 | NP_001239415.1 | 13 | 1714 | |||
NM_133247.3 | NP_573510.2 | 13 | 1714 | |||
GenBank mRNA | AK082251.1 | - | 13 | 1697 | ||
AK082943.1 | - | 13 | 1697 | |||
AK087467.1 | - | 13 | 1715 | |||
AK143844.1 | - | 13 | 1682 | |||
BC031366.1 | - | 5 | 830 | |||
BC036980.1 | - | 6 | 914 | |||
BC067399.1 | - | 12 | 1556 | |||
BC089315.1 | - | 13 | 1660 | |||
CN718979.1 | - | 2 | 173 |