Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Usp33 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001076676.2 | NP_001070144.1 | 14 | 1794 | ||
NM_001252486.1 | NP_001239415.1 | 14 | 1794 | |||
NM_133247.3 | NP_573510.2 | 14 | 1818 | |||
GenBank mRNA | AF383174.1 | - | 14 | 1807 | ||
AK082251.1 | - | 14 | 1777 | |||
AK082943.1 | - | 14 | 1777 | |||
AK087467.1 | - | 14 | 1795 | |||
AK122442.1 | - | 12 | 3021 | |||
AK143844.1 | - | 14 | 1786 | |||
BC005506.1 | - | 14 | 1747 | |||
BC031366.1 | - | 6 | 934 | |||
BC036980.1 | - | 7 | 994 | |||
BC049870.1 | - | 1 | 59 | |||
BC067399.1 | - | 13 | 1660 | |||
BC089315.1 | - | 14 | 1740 | |||
CN718979.1 | - | 3 | 253 |