Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Glmn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001161738.1 | NP_001155210.1 | 9 | 1063 | ||
NM_001161739.1 | NP_001155211.1 | 9 | 1077 | |||
NM_133248.2 | NP_573511.2 | 9 | 1077 | |||
GenBank mRNA | AJ566083.1 | - | 8 | 945 | ||
AK034160.1 | - | 9 | 1064 | |||
AK165661.1 | - | 9 | 1048 | |||
AK169045.1 | - | 9 | 1049 | |||
AK172659.1 | - | 9 | 1015 | |||
BC003446.1 | - | 9 | 1013 | 2651614 |