Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ppargc1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133249.2 | NP_573512.1 | 11, 12 | 3000 | ||
GenBank mRNA | AF453324.1 | - | 11, 12 | 3000 | ||
AK028464.1 | - | 12, 13 | 3050 | |||
AK086111.1 | - | 11, 12 | 3023 | |||
BC150699.1 | - | 11, 12 | 2980 | 9087610 |