Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PBX3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001134778.1 | NP_001128250.1 | 8 | 1169 | ||
NM_006195.5 | NP_006186.1 | 8 | 1255 | |||
NR_024122.1 | 7 | 1013 | ||||
NR_024123.1 | 6 | 900 | ||||
XM_006717130.1 | XP_006717193.1 | 9 | 1284 | |||
XM_006717131.1 | XP_006717194.1 | 8 | 1171 | |||
XM_006717132.1 | XP_006717195.1 | 7 | 1108 | |||
XM_006717133.1 | XP_006717196.1 | 8 | 945 | |||
XM_011518755.1 | XP_011517057.1 | 9 | 1232 | |||
XM_017014786.1 | XP_016870275.1 | 7 | 1056 | |||
GenBank mRNA | AF355141.1 | - | 7 | 897 | ||
AF355142.1 | - | 6 | 784 | |||
AK289376.1 | - | 7 | 882 | |||
AK297093.1 | - | 8 | 1124 | |||
AK299267.1 | - | 6 | 852 | |||
AK309713.1 | - | 8 | 1169 | |||
AK312717.1 | - | 8 | 1185 | |||
BC016977.1 | - | 7 | 938 | |||
BC094883.1 | - | 8 | 1206 | 5212090 | ||
X59841.1 | - | 8 | 1251 |