Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Krtap19-9b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133359.2 | NP_579937.1 | 1 | 286 | ||
GenBank mRNA | AK086935.1 | - | 1 | 288 | ||
AY026312.1 | - | 1 | 287 | |||
BC119613.1 | - | 1 | 275 | 40061693 | ||
BC119614.1 | - | 1 | 275 | 40061695 | ||
BC127239.1 | - | 1 | 275 | |||
BC127240.1 | - | 1 | 275 |
Gene Symbol: Krtap19-9a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AF477980.1 | - | 260 |