Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Acer1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175731.4 | NP_783858.1 | 6 | 697 | ||
GenBank mRNA | AF347023.1 | - | 6 | 679 | ||
AK028901.1 | - | 6 | 698 | |||
AK075884.1 | - | 6 | 683 | |||
BC130254.1 | - | 6 | 702 | 40142524 |