Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000920.3 | NP_000911.2 | 14 | 1853 | ||
NM_001040716.1 | NP_001035806.1 | 15 | 2041 | |||
NM_022172.2 | NP_071504.2 | 13 | 1808 | |||
XM_005274031.4 | XP_005274088.1 | 13 | 1900 | |||
XM_005274032.4 | XP_005274089.1 | 13 | 1782 | |||
XM_006718578.3 | XP_006718641.1 | 13 | 1853 | |||
XM_006718579.3 | XP_006718642.1 | 5 | 642 | |||
XM_011545086.2 | XP_011543388.1 | 13 | 2169 | |||
XM_011545087.2 | XP_011543389.1 | 5 | 767 | |||
XM_017017868.1 | XP_016873357.1 | 16 | 2340 | |||
XM_017017869.1 | XP_016873358.1 | 16 | 2211 | |||
XM_017017870.1 | XP_016873359.1 | 15 | 2152 | |||
XM_017017871.1 | XP_016873360.1 | 15 | 2023 | |||
XM_017017872.1 | XP_016873361.1 | 14 | 2405 | |||
GenBank mRNA | BC011617.2 | - | 14 | 1842 | 3946441 | |
S72370.1 | - | 13 | 1841 | |||
U04641.1 | - | 13 | 1797 | |||
U30891.1 | - | 14 | 1811 |