Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Acer1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175731.4 | NP_783858.1 | 4, 5 | 525 | ||
GenBank mRNA | AF347023.1 | - | 4, 5 | 507 | ||
AK028901.1 | - | 4, 5 | 526 | |||
AK075884.1 | - | 4, 5 | 511 | |||
BC130254.1 | - | 4, 5 | 530 | 40142524 |