Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000920.3 | NP_000911.2 | 8 | 866 | ||
NM_001040716.1 | NP_001035806.1 | 9 | 1054 | |||
NM_022172.2 | NP_071504.2 | 7 | 821 | |||
XM_005274031.4 | XP_005274088.1 | 7 | 913 | |||
XM_005274032.4 | XP_005274089.1 | 7 | 795 | |||
XM_006718578.3 | XP_006718641.1 | 7 | 866 | |||
XM_011545086.2 | XP_011543388.1 | 7 | 1182 | |||
XM_017017868.1 | XP_016873357.1 | 10 | 1353 | |||
XM_017017869.1 | XP_016873358.1 | 10 | 1224 | |||
XM_017017870.1 | XP_016873359.1 | 9 | 1165 | |||
XM_017017871.1 | XP_016873360.1 | 9 | 1036 | |||
XM_017017872.1 | XP_016873361.1 | 8 | 1418 | |||
GenBank mRNA | AK297705.1 | - | 8 | 866 | ||
BC011617.2 | - | 8 | 855 | 3946441 | ||
S72370.1 | - | 7 | 854 | |||
U04641.1 | - | 7 | 810 | |||
U30891.1 | - | 8 | 824 |