Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000920.3 | NP_000911.2 | 4, 5 | 423 | ||
NM_001040716.1 | NP_001035806.1 | 5, 6 | 611 | |||
NM_022172.2 | NP_071504.2 | 3, 4 | 378 | |||
XM_005274031.4 | XP_005274088.1 | 3, 4 | 470 | |||
XM_005274032.4 | XP_005274089.1 | 3, 4 | 352 | |||
XM_006718578.3 | XP_006718641.1 | 3, 4 | 423 | |||
XM_011545086.2 | XP_011543388.1 | 3, 4 | 739 | |||
XM_017017868.1 | XP_016873357.1 | 6, 7 | 910 | |||
XM_017017869.1 | XP_016873358.1 | 6, 7 | 781 | |||
XM_017017870.1 | XP_016873359.1 | 5, 6 | 722 | |||
XM_017017871.1 | XP_016873360.1 | 5, 6 | 593 | |||
XM_017017872.1 | XP_016873361.1 | 4, 5 | 975 | |||
GenBank mRNA | AK297705.1 | - | 4, 5 | 423 | ||
BC011617.2 | - | 4, 5 | 412 | 3946441 | ||
CN267865.1 | - | 5, 6 | 616 | |||
DA542083.1 | - | 4, 5 | 423 | |||
S72370.1 | - | 3, 4 | 411 | |||
U04641.1 | - | 3, 4 | 367 | |||
U30891.1 | - | 4, 5 | 381 |