Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Vmn1r32 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_134170.3 | NP_598931.1 | 3 | 2121 | ||
GenBank mRNA | AK029205.1 | - | 3 | 2122 | ||
BC119596.1 | - | 1 | 1784 | 40059150 | ||
BC119597.1 | - | 1 | 1784 | 40059151 |