Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Vmn1r4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_134176.1 | NP_598937.1 | 1 | 501 | ||
GenBank mRNA | AK082576.1 | - | 5 | 1151 | ||
BC132386.1 | - | 1 | 548 | 40130663 | ||
BC138489.1 | - | 1 | 548 | 8861509 |