Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PCBD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000281.3 | NP_000272.1 | 2 | 366 | ||
NM_001289797.1 | NP_001276726.1 | 2 | 512 | |||
NM_001323004.1 | NP_001309933.1 | 2 | 366 | |||
GenBank mRNA | AA397961.1 | - | 1 | 127 | ||
AF082858.1 | - | 2 | 186 | |||
BC006324.2 | - | 2 | 131 | 4138350 | ||
BI548743.1 | - | 2 | 517 | |||
BM724638.1 | - | 1 | 76 | |||
BQ685076.1 | - | 364 | ||||
BQ897536.1 | - | 2 | 396 | |||
CR542194.1 | - | 1 | 113 | |||
L41559.1 | - | 2 | 133 | |||
M83742.1 | - | 1 | 114 |