Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Vmn1r28 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_134180.1 | NP_598941.1 | 1 | 177 | ||
GenBank mRNA | AF394949.1 | - | 333 |
Gene Symbol: Vmn1r18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_134181.1 | NP_598942.1 | 1 | 177 | ||
GenBank mRNA | BC132061.1 | - | 1 | 276 | 40130338 | |
BC138409.1 | - | 1 | 276 | 8861429 |
Gene Symbol: Vmn1r19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_134182.1 | NP_598943.1 | 1 | 177 | ||
GenBank mRNA | BC125351.1 | - | 1 | 275 | 40129866 | |
BC132235.1 | - | 1 | 275 | 40130512 |