Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Vmn1r8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_134187.3 | NP_598948.1 | 1 | 598 | ||
GenBank mRNA | AF394951.1 | - | 1 | 598 |
Gene Symbol: Vmn1r7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001166710.1 | NP_001160182.1 | 1 | 541 |