Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Arhgap4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001162423.1 | NP_001155895.1 | 6 | 829 | ||
NM_001162424.1 | NP_001155896.1 | 5 | 766 | |||
NM_138630.2 | NP_619536.2 | 6 | 829 | |||
GenBank mRNA | AF443826.1 | - | 6 | 760 | ||
AK129060.1 | - | 1244 | ||||
AK157294.1 | - | 6 | 828 | |||
AY183124.1 | - | 6 | 802 | |||
AY183125.1 | - | 5 | 739 | |||
AY183126.1 | - | 6 | 802 | |||
AY183127.1 | - | 6 | 802 | |||
AY183129.1 | - | 6 | 802 | |||
AY183133.1 | - | 5 | 739 | |||
BC109001.1 | - | 6 | 760 | 40048081 | ||
BC109002.1 | - | 5 | 697 |